Tetrabromomethane, (max. 6% H₂O) 98% (dry weight)

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

L00821.14 L00821.22 L00821.36
L00821.14EA 35.8 EUR
L00821.14 L00821.22 L00821.36
Tetrabromomethane, (max. 6% H₂O) 98% (dry weight)
Tetrabromomethane

Purity: 98% CAS No.: 558-13-4 Molecular Formula: CBr4 Formula Weight: 331.63 Melting Point: 90-95° Boiling Point: 190° UN No.: 2516 MDL No.: MFCD00000117

Formule: Br₄C
Kookpunt: 190 °C
Smeltpunt: 90…95 °C
Dichtheid: 3,420
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00000117
CAS nummer: 558-13-4
EINECS: 209-189-6
UN: 2516
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR