Cell culture media, DMEM-Ham's F12
Leverancier: BIOWEST
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
P0095-N1L P0095-N10L L0090-500 L0092-500 L0093-500 L0094-500 L0095-500 L0096-500 L0091-500
P0095-N1LEA 25.9 EUR
P0095-N1L P0095-N10L L0090-500 L0092-500 L0093-500 L0094-500 L0095-500 L0096-500 L0091-500
Cell culture media, DMEM-Ham's F12
Celcultuurmedia
Studies to determine the nutritional requirements of many cells have been in progress since Eagle's first reports. The major essential nutrients were identified, and work became focussed on the media requirements of individual cell types. Many media designed for these purposes are now available. Among the first of these media, developed initially to study hormonal requirements of cells in culture, was a mixture of DMEM and Ham's F12 medium, known as DMEM-Ham's F12.