6-Amino-1,4-benzodioxane 99%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

L01421.06 L01421.14 L01421.22
L01421.06EA 29.3 EUR
L01421.06 L01421.14 L01421.22
6-Amino-1,4-benzodioxane 99%
6-Amino-1,4-benzodioxane

Purity: 99% CAS No.: 22013-33-8 Molecular Formula: C8H9NO2 Formula Weight: 151.17 Flash Point: 110°(230°F) Refractive Index: 1.5990 Melting Point: 26-30° Boiling Point: 116-118°/3mm Beilstein Reference: 6447 UN No.: 2811 MDL No.: MFCD00006824

Formule: C₈H₉NO₂
Kookpunt: 116…118 °C (3 mmHg)
Smeltpunt: 26…30 °C
Dichtheid: 1,231
Vlampunt: >110 °C (230 °F)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00006824
CAS nummer: 22013-33-8
EINECS: 244-718-4
UN: 2811
ADR: 6.1,III

Order Now

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR