Benzylbenzoaat ≥99%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

Synoniemen: Benzoic acid benzyl ester

L03258.30 L03258.0B L03258.0E
L03258.30EA 25.5 EUR
L03258.30 L03258.0B L03258.0E
Benzylbenzoaat ≥99%
Benzylbenzoaat

Purity: 99+% CAS No.: 120-51-4 Molecular Formula: C14H12O2 Formula Weight: 212.25 Flash Point: 147°(296°F) Refractive Index: 1.5680 Melting Point: 18-20° Boiling Point: 323-324° Beilstein Reference: 2049280 Merck Reference: 14,1127 UN No.: 3082 MDL No.: MFCD00003075

Formule: C₁₄H₁₂O₂
MW: 212.25 g/mol
Kookpunt: 323…324 °C
Smeltpunt: 18…20 °C
Dichtheid: 1.112
Vlampunt: 147 °C (296 °F)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00003075
CAS nummer: 120-51-4
EINECS: 204-402-9
UN: 3082
ADR: 9 III
Merck Index: 13,01129

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR