1,5-Dihydroxynaphthalene ≥98%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

L04599.30 L04599.0B
L04599.30EA 65.9 EUR
L04599.30 L04599.0B
1,5-Dihydroxynaphthalene ≥98%
1,5-Dihydroxynaphthalene

Purity: 98% CAS No.: 83-56-7 Molecular Formula: C10H8O2 Formula Weight: 160.17 Flash Point: 252°(486°F) Melting Point: ca 260° dec. Beilstein Reference: 2044951 MDL No.: MFCD00003980

Formule: C₁₀H₈O₂
Kookpunt: 375 °C (1013 hPa)
Dichtheid: 1,41 g/cm³ (20 °C)
Vlampunt: 252 °C (486 °F)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00003980
CAS nummer: 83-56-7
EINECS: 201-487-4
UN: 3077
ADR: 9,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR