n-Butyl phenyl ether ≥99%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

L04703.14 L04703.22
L04703.14EA 48 EUR
L04703.14 L04703.22
n-Butyl phenyl ether ≥99%
n-Butyl phenyl ether

Purity: 99% CAS No.: 1126-79-0 Molecular Formula: C6H5OCH2CH2CH2CH3 Formula Weight: 150.22 Flash Point: 82°(179°F) Refractive Index: 1.4970 Melting Point: -19° Boiling Point: 210-211° Beilstein Reference: 1635559 MDL No.: MFCD00009438

Formule: C₁₀H₁₄O
Kookpunt: 210…211 °C
Smeltpunt: –19 °C
Dichtheid: 0,938
Vlampunt: 82 °C (179 °F)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00009438
CAS nummer: 1126-79-0
EINECS: 214-426-1

Order Now

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR