1-(2-Pyridyl)piperazine ≥99%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

L05451.06 L05451.14 L05451.22
L05451.06EA 34.7 EUR
L05451.06 L05451.14 L05451.22
1-(2-Pyridyl)piperazine ≥99%
1-(2-Pyridyl)piperazine

Purity: 99% CAS No.: 34803-66-2 Molecular Formula: C9H13N3 Formula Weight: 163.22 Flash Point: 110°(230°F) Refractive Index: 1.5950 Boiling Point: 120-122°/2mm Beilstein Reference: 140423 UN No.: 2735 MDL No.: MFCD00006216

Formule: C₉H₁₃N₃
Kookpunt: 120…122 °C (2 mmHg)
Smeltpunt: 85…87 °C
Dichtheid: 1,072
Vlampunt: >110 °C (230 °F)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00006216
CAS nummer: 34803-66-2
EINECS: 252-220-3
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR