(±)-2-Decanol ≥98%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

L05688.06 L05688.14
L05688.06EA 29.2 EUR
L05688.06 L05688.14
(±)-2-Decanol ≥98%
(±)-2-Decanol

Purity: 98% CAS No.: 1120-06-5 Molecular Formula: C10H22O Formula Weight: 158.29 Flash Point: 85°(185°F) Refractive Index: 1.4340 Melting Point: -5° Boiling Point: 211° Beilstein Reference: 1719664 MDL No.: MFCD00004594

Formule: C₁₀H₂₂O
Kookpunt: 211 °C
Smeltpunt: –5 °C
Dichtheid: 0,825
Vlampunt: 85 °C (185 °F)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00004594
CAS nummer: 1120-06-5
EINECS: 214-296-6

Order Now

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR