1-Methylpyrrole-2-carboxylic acid ≥98%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

L05906.03 L05906.06
L05906.03EA 26.7 EUR
L05906.03 L05906.06
1-Methylpyrrole-2-carboxylic acid ≥98%
1-Methylpyrrole-2-carboxylic acid

Purity: 98% CAS No.: 6973-60-0 Molecular Formula: C6H7NO2 Formula Weight: 125.13 Melting Point: 135-138° Beilstein Reference: 115703

Formule: C₆H₇NO₂
Smeltpunt: 135…138 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00003088
CAS nummer: 6973-60-0
EINECS: 230-217-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR