Dabsyl chloride ≥98%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

Synoniemen: DABS-Cl, 4-Dimethylaminoazobenzene-4'-sulphonyl Chloride, 4-((4-(Dimethylamino)phenyl)diazenyl)benzenesulphonyl chloride

L06057.MCEA 28.8 EUR
L06057.MC L06057.ME
Dabsyl chloride ≥98%
Dabsyl chloride

Purity: 98+% CAS No.: 56512-49-3 Molecular Formula: C14H14ClN3O2S Formula Weight: 323.80 Storage Temperature: 2°C to 8°C Melting Point: ca 185° dec. Beilstein Reference: 3064095 UN No.: UN3261 MDL No.: MFCD00007444

Formule: C₁₄H₁₄ClN₃O₂S
MW: 323.80 g/mol
Smeltpunt: ∼ 185 °C (ontbinding)
MDL Nummer: MFCD00007444
CAS nummer: 56512-49-3
EINECS: 260-235-1
ADR: 8 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR