3-Pentanol ≥98%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

L06065.14 L06065.22 L06065.36
L06065.14EA 28.5 EUR
L06065.14 L06065.22 L06065.36
3-Pentanol ≥98%
3-Pentanol

Purity: 98+% CAS No.: 584-02-1 Molecular Formula: C5H12O Formula Weight: 88.15 Flash Point: 33°(91°F) Refractive Index: 1.4100 Odour: Characteristic Melting Point: -8° Boiling Point: 114-115° Beilstein Reference: 1730964 Merck Reference: 14,7120 UN No.: 1105 MDL No.: MFCD00004574

Formule: C₅H₁₂O
Kookpunt: 114…115 °C
Smeltpunt: –8 °C
Dichtheid: 0,817
Vlampunt: 33 °C (91 °F)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00004574
CAS nummer: 584-02-1
EINECS: 209-526-7
UN: 1105
ADR: 3,III
Merck Index: 14,7120
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2ec7192926b50615dfac930445c63174c6221a44&vid=a2f1fa3f1ffa1ecbfb6a5dd1d0f29da27d1d8737&prid=web&clid=SAPDAM

Order Now

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR