Cell culture salts, salt solutions and buffers
Leverancier: BIOWEST
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
L0180-100 L0180-500 L0601-500 L0607-500 L0611-500 L0615-100 L0615-500 X0113-500 X0510-500 X0515-500 X0520-500 L0640-500 L0642-100 L0643-100 L0680-100 P0750-N1L P0750-N10L P0750-N5L X0112-500 L0642-500 L0680-500 P0750-N50L P0153-N1L P0154-N10L P0154-N1L P0154-N50L P5455-100GR P2064-N5L P2035-500GR P0153-N10L P2060-500GR L0606-500 L0643-500 L0611-100 L0612-500 L0615-1000 L0605-500 X0507-500 X0112-100 X0509-500 X0513-500 X0515-100 L0615-C10LS P2060-1KG P2066-1KG L0606-100
L0180-100EA 26.6 EUR
L0180-100 L0180-500 L0601-500 L0607-500 L0611-500 L0615-100 L0615-500 X0113-500 X0510-500 X0515-500 X0520-500 L0640-500 L0642-100 L0643-100 L0680-100 P0750-N1L P0750-N10L P0750-N5L X0112-500 L0642-500 L0680-500 P0750-N50L P0153-N1L P0154-N10L P0154-N1L P0154-N50L P5455-100GR P2064-N5L P2035-500GR P0153-N10L P2060-500GR L0606-500 L0643-500 L0611-100 L0612-500 L0615-1000 L0605-500 X0507-500 X0112-100 X0509-500 X0513-500 X0515-100 L0615-C10LS P2060-1KG P2066-1KG L0606-100
Cell culture salts, salt solutions and buffers
Buffers Buffers voor weefselkweek
These irrigating buffers and salt solutions are sterile, physiological balanced solutions intended for use in the maintenance of mammalian cells, where a chemically defined, balanced salt solution provides an environment that will maintain the structural and physiological integrity of cells in vitro. Applications include the intermediate steps for cultivation of cells, like washing, centrifugation, suspending and counting, as well as for many analytical methods or biochemical treatments. Principally these solutions should be used to keep the cells in suspension only for minutes or a few hours maximum. These buffers and solutions are not cell culture media.
They are comprised of a phosphate buffer system, sodium chloride to adjust the osmolarity and perhaps sugar for short term nutrition and stabilisation of morphology. For applications where Ca²⁺ and Mg²⁺ ions interfere with enzyme activity (eg. Trypsin) the modified buffers without calcium and magnesium should be used.
Order Now
Specificaties
Environmentally Preferable