Cyclohexanemethylamine 98%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

Synoniemen: (Cyclohexylmethyl)amine, (Aminomethyl)cyclohexane, Cyclohexylmethanamine

L06501.06 L06501.14
L06501.06EA 38.4 EUR
L06501.06 L06501.14
Cyclohexanemethylamine 98%
Cyclohexanemethylamine

Purity: 98% CAS No.: 3218-02-8 Molecular Formula: C7H15N Formula Weight: 113.20 Flash Point: 43°(109°F) Refractive Index: 1.4620 Melting Point: -8° Boiling Point: 161-163° Beilstein Reference: 635751 UN No.: 2734 MDL No.: MFCD00001520

Formule: C₇H₁₅N
MW: 113.2 g/mol
Kookpunt: 161…163 °C
Smeltpunt: –8 °C
Dichtheid: 0.871
Vlampunt: 43 °C (109 °F)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00001520
CAS nummer: 3218-02-8
EINECS: 221-741-8
UN: 2734
ADR: 8 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR