2-Furoyl chloride ≥98%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

L06960.18 L06960.30
L06960.18EA 56.1 EUR
L06960.18 L06960.30
2-Furoyl chloride ≥98%
2-Furoyl chloride

Purity: 98+% CAS No.: 527-69-5 Molecular Formula: C5H3ClO2 Formula Weight: 130.53 Flash Point: 85°(185°F) Refractive Index: 1.5310 Melting Point: -2 to 1° Boiling Point: 173-174° Beilstein Reference: 110144 Merck Reference: 14,4310 UN No.: 3265 MDL No.: MFCD00003228

Formule: C₅H₃ClO₂
Kookpunt: 173…174 °C
Smeltpunt: –2…+1 °C
Dichtheid: 1,325
Vlampunt: 85 °C (185 °F)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00003228
CAS nummer: 527-69-5
EINECS: 208-422-9
UN: 3265
ADR: 8,II
Merck Index: 14,4310

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR