D(-)-Lysine monohydrochloride ≥98%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synoniemen: (R)-(-)-Lysine monohydrochloride, H-D-Lys-OH.HCl, D-Lysine hydrochloride, (R)-(-)-2,6-Diaminocaproic acid monohydrochloride, (R)-(-)-2,6-Diaminohexanoic acid monohydrochloride

L07710.03 L07710.06
L07710.03EA 35.5 EUR
L07710.03 L07710.06
D(-)-Lysine monohydrochloride ≥98%
D(-)-Lysine monohydrochloride

Purity: 98% CAS No.: 7274-88-6 Molecular Formula: C6H14N2O2.HCl Formula Weight: 182.65 Melting Point: ca 260° dec. Beilstein Reference: 4356907

Formule: C₆H₁₅ClN₂O₂
MW: 182.65 g/mol
Smeltpunt: ∼ 260 °C (ontbinding)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00012920
CAS nummer: 7274-88-6
EINECS: 230-691-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR