2,5-Dichlorobenzyl alcohol ≥99%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

L07734.06 L07734.14
L07734.06EA 38.6 EUR
L07734.06 L07734.14
2,5-Dichlorobenzyl alcohol ≥99%
2,5-Dichlorobenzyl alcohol

Purity: 99% CAS No.: 34145-05-6 Molecular Formula: C7H6Cl2O Formula Weight: 177.03 Melting Point: 76-80° Beilstein Reference: 3240770

Formule: C₇H₆Cl₂O
Kookpunt: 264 °C (1013 hPa)
Smeltpunt: 76…80 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00004607
CAS nummer: 34145-05-6
EINECS: 251-850-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR