2-Butanonoxim ≥99%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

L08196.AE L08196.AP L08196.AU
L08196.AEEA 27.2 EUR
L08196.AE L08196.AP L08196.AU
2-Butanonoxim ≥99%
2-Butanonoxim

Purity: 99% CAS No.: 96-29-7 Molecular Formula: C4H9NO Formula Weight: 87.12 Flash Point: 62°(144°F) Refractive Index: 1.4430 Melting Point: -30° Boiling Point: 152-153° Beilstein Reference: 1698241 MDL No.: MFCD00013935

Formule: C₄H₉NO
Kookpunt: 152…153 °C
Smeltpunt: –30 °C
Dichtheid: 0,921
Vlampunt: 62 °C (144 °F)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00013935
CAS nummer: 96-29-7
EINECS: 202-496-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR