Hexyltrimethylammonium bromide ≥98%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

L08594.06 L08594.14
L08594.06EA 38.3 EUR
L08594.06 L08594.14
Hexyltrimethylammonium bromide ≥98%
Hexyltrimethylammonium bromide

Purity: 98% CAS No.: 2650-53-5 Molecular Formula: CH3(CH2)5N(CH3)3Br Formula Weight: 224.18 Melting Point: 177-180° Beilstein Reference: 3913229

Formule: C₉H₂₂BrN
Smeltpunt: 177…180 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00051872
CAS nummer: 2650-53-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR