4-Bromobenzenesulphonamide ≥98%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

L08602.03 L08602.06 L08602.14
L08602.03EA 29.2 EUR
L08602.03 L08602.06 L08602.14
4-Bromobenzenesulphonamide ≥98%
4-Bromobenzenesulphonamide

Purity: 98% CAS No.: 701-34-8 Molecular Formula: C6H6BrNO2S Formula Weight: 236.09 Melting Point: 164-167° Beilstein Reference: 2691657

Formule: C₆H₆BrNO₂S
Smeltpunt: 164…167 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00051977
CAS nummer: 701-34-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR