2,4-Dimethoxyphenyl isocyanate ≥97%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

Synoniemen: 1-Isocyanato-2,4-dimethoxybenzene

L09762.06 L09762.03
L09762.06EA 79.6 EUR
L09762.06 L09762.03
2,4-Dimethoxyphenyl isocyanate ≥97%
2,4-Dimethoxyphenyl isocyanate

Purity: 97% CAS No.: 84370-87-6 Molecular Formula: C9H9NO3 Formula Weight: 179.18 Storage Temperature: 2°C to 8°C Flash Point: 110°(230°F) Refractive Index: 1.5475 Melting Point: 31-32° Boiling Point: 139-141°/11mm Beilstein Reference: 1106649 UN No.: 2811 MDL No.: MFCD00002005

Formule: C₉H₉NO₃
Kookpunt: 139…141 °C (11 mmHg)
Smeltpunt: 31…32 °C
Vlampunt: >110 °C (230 °F)
MDL Nummer: MFCD00002005
CAS nummer: 84370-87-6
EINECS: 282-747-4
UN: 2811
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR