(Aminomethyl)phosphonic acid 99%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

L09833.03 L09833.MD L09833.06
L09833.03EA 142 EUR
L09833.03 L09833.MD L09833.06
(Aminomethyl)phosphonic acid 99%
(Aminomethyl)phosphonic acid

Purity: 99% CAS No.: 1066-51-9 Molecular Formula: CH6NO3P Formula Weight: 111.04 Melting Point: ca 290° dec. Beilstein Reference: 1701219

Formule: CH₆NO₃P
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00008105
CAS nummer: 1066-51-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR