2-Tetrahydrofurfuryl isothiocyanate 98%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

L09878.06 L09878.03
L09878.06EA 122 EUR
L09878.06 L09878.03
2-Tetrahydrofurfuryl isothiocyanate 98%
2-Tetrahydrofurfuryl isothiocyanate

Purity: 98% CAS No.: 36810-87-4 Molecular Formula: C6H9NOS Formula Weight: 143.21 Storage Temperature: 2°C to 8°C Refractive Index: 1.5350 Boiling Point: 122-125°/18mm Beilstein Reference: 111696 UN No.: UN2922 MDL No.: MFCD00022476

Formule: C₆H₉NOS
Kookpunt: 122…125 °C (18 mmHg)
Dichtheid: 1,120
MDL Nummer: MFCD00022476
CAS nummer: 36810-87-4
ADR: 8,III

Order Now

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR