Cyclohexene-1-carbonitrile ≥98%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

L10284.09 L10284.04
L10284.09EA 178 EUR
L10284.09 L10284.04
Cyclohexene-1-carbonitrile ≥98%
Cyclohexene-1-carbonitrile

Purity: 98% CAS No.: 1855-63-6 Molecular Formula: C7H9N Formula Weight: 107.16 Flash Point: 83°(181°F) Refractive Index: 1.4820 Boiling Point: 85-88°/17mm Beilstein Reference: 385837 UN No.: 3276 MDL No.: MFCD00013772

Formule: C₇H₉N
Kookpunt: 85…88 °C (17 mmHg)
Dichtheid: 0,947
Vlampunt: 83 °C (181 °F)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00013772
CAS nummer: 1855-63-6
EINECS: 217-454-2
UN: 3276
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR