Hydriodinezuur 57% (w/w) in aqueous solution stabilised

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

Synoniemen: Hydrogeenjodide

Stabilisatie: stab. with hypophosphorous acid 1,5%

L10410.22 L10410.36
L10410.22EA 38.5 EUR
L10410.22 L10410.36
Hydriodinezuur 57% (w/w) in aqueous solution stabilised
Hydriodic acid

CAS No.: 10034-85-2 Molecular Formula: HI Formula Weight: 127.91 Physical Form: Liquid Density: 1.701 Sensitivity: Light Sensitive Appearance: Colourless to yellow Odour: Pungent Boiling Point: 126-127° Merck Reference: 14,4776 UN No.: 1787 MDL No.: MFCD00011347

Formule: HI
Kookpunt: 126…127 °C
Dichtheid: 1,701
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00011347
CAS nummer: 10034-85-2
EINECS: 233-109-9
UN: 1787
ADR: 8,II
Merck Index: 14,4776

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR