Triethyl-4-phosphonobutyrate ≥97%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

L10488.09 L10488.04
L10488.09EA 117 EUR
L10488.09 L10488.04
Triethyl-4-phosphonobutyrate ≥97%
Triethyl-4-phosphonobutyrate

Purity: 97% CAS No.: 2327-69-7 Molecular Formula: C10H21O5P Formula Weight: 252.25 Refractive Index: 1.4370 Boiling Point: 117-119°/0.4mm MDL No.: MFCD00015168

Formule: C₁₀H₂₁O₅P
Kookpunt: 117…119 °C (0,4 mmHg)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00015168
CAS nummer: 2327-69-7

Order Now

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR