Dimethyl adipimidate dihydrochloride 97%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

L10515.09 L10515.04
L10515.09EA 151 EUR
L10515.09 L10515.04
Dimethyl adipimidate dihydrochloride 97%
Dimethyl adipimidate dihydrochloride

Purity: 97% CAS No.: 14620-72-5 Molecular Formula: C8H16N2O2.2HCl Formula Weight: 245.15 Melting Point: ca 215° dec. MDL No.: MFCD00012573

Formule: C₈H₁₆N₂O₂
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00012573
CAS nummer: 14620-72-5
EINECS: 238-660-9

Order Now

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR