2-Isopropylphenyl isocyanate ≥97%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

L10618.03 L10618.06 L10618.14
L10618.03EA 26.2 EUR
L10618.03 L10618.06 L10618.14
2-Isopropylphenyl isocyanate ≥97%
2-Isopropylphenyl isocyanate

Purity: 97% CAS No.: 56309-56-9 Molecular Formula: C10H11NO Formula Weight: 161.21 Storage Temperature: 2°C to 8°C Flash Point: 87°(188°F) Refractive Index: 1.5230 Boiling Point: 66-67°/5mm Beilstein Reference: 3588928 UN No.: 2206 MDL No.: MFCD00013850

Formule: C₁₀H₁₁NO
Kookpunt: 66…67 °C (5 mmHg)
Dichtheid: 1,019
Vlampunt: 87 °C (188 °F)
MDL Nummer: MFCD00013850
CAS nummer: 56309-56-9
EINECS: 260-105-4
ADR: 6.1,II

Order Now

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR