Cyclohexyl methacrylate 97% stabilised

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing


Stabilisatie: Stabilised with 4-methoxyphenol (MEHQ) 50 ppm

L10634.22
L10634.22EA 56.7 EUR
L10634.22
Cyclohexyl methacrylate 97% stabilised
Cyclohexyl methacrylate

Stab. with ca 50ppm 4-methoxyphenol


Purity: 97% CAS No.: 101-43-9 Molecular Formula: C10H16O2 Formula Weight: 168.24 Flash Point: 82°(179°F) Refractive Index: 1.4577 Boiling Point: 72-73°/5mm Beilstein Reference: 2045235 MDL No.: MFCD00014292

Formule: C₁₀H₁₆O₂
Kookpunt: 72…73 °C (5 mmHg)
Dichtheid: 0,970
Vlampunt: 82 °C (179 °F)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00014292
CAS nummer: 101-43-9
EINECS: 202-943-5

Order Now

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR