(±)-2,4-Difluoromandelic acid ≥97%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

L10708.03 L10708.06
L10708.03EA 63.8 EUR
L10708.03 L10708.06
(±)-2,4-Difluoromandelic acid ≥97%
(±)-2,4-Difluoromandelic acid

Purity: 97% CAS No.: 132741-30-1 Molecular Formula: C8H6F2O3 Formula Weight: 188.13 Melting Point: 122-124° MDL No.: MFCD00061296

Formule: C₈H₆F₂O₃
Smeltpunt: 122…124 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00061296
CAS nummer: 132741-30-1

Order Now

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR