Methyl-3,5-diaminobenzoate ≥99%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

L10848.09 L10848.18
L10848.09EA 62.8 EUR
L10848.09 L10848.18
Methyl-3,5-diaminobenzoate ≥99%
Methyl-3,5-diaminobenzoate

Purity: 99% CAS No.: 1949-55-9 Molecular Formula: C8H10N2O2 Formula Weight: 166.18 Odour: Odourless Melting Point: 131-134° Beilstein Reference: 2363309 MDL No.: MFCD00041433

Formule: C₈H₁₀N₂O₂
Kookpunt: 319 °C (1013 hPa)
Smeltpunt: 131…134 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00041433
CAS nummer: 1949-55-9
EINECS: 000-000-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR