4-(n-Butoxy)benzenesulphonyl chloride 97%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

Synoniemen: 4-Butoxybenzenesulphonyl chloride, 4-Butoxybenzene-1-sulphonyl chloride, 4-Butoxyphenylsulphonyl chloride

L11258.03 L11258.06
L11258.03EA 61.9 EUR
L11258.03 L11258.06
4-(n-Butoxy)benzenesulphonyl chloride 97%
4-(n-Butoxy)benzenesulphonyl chloride

Purity: 97% CAS No.: 1138-56-3 Molecular Formula: C10H13ClO3S Formula Weight: 248.73 Refractive Index: 1.5420 Boiling Point: 130-131°/0.6mm Beilstein Reference: 1109264 UN No.: UN3265 MDL No.: MFCD00052344

Formule: C₁₀H₁₃ClO₃S
MW: 248.73 g/mol
Kookpunt: 130…131 °C (0.6 mmHg)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00052344
CAS nummer: 1138-56-3
ADR: 8 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR