Dithiodiglycol (bis(2-hydroxyethyl) disulphide) tech. 90%, Technical Grade

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

L11800.18 L11800.30
L11800.18EA 64.6 EUR
L11800.18 L11800.30
Dithiodiglycol (bis(2-hydroxyethyl) disulphide) tech. 90%, Technical Grade
Dithiodiglycol (bis(2-hydroxyethyl) disulphide)

Purity: 90% CAS No.: 1892-29-1 Molecular Formula: C4H10O2S2 Formula Weight: 154.24 Flash Point: 110°(230°F) Refractive Index: 1.5700 Odour: Unpleasant Melting Point: ca 25° Boiling Point: 158-163°/3.5mm Beilstein Reference: 1735851 UN No.: 2810 MDL No.: MFCD00002906

Formule: C₄H₁₀O₂S₂
Kookpunt: 158…163 °C (3,5 mmHg)
Smeltpunt: ∼ 25 °C
Dichtheid: 1,261
Vlampunt: >110 °C (230 °F)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00002906
CAS nummer: 1892-29-1
EINECS: 217-576-6
UN: 2810
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR