3,4-Dichlorothiophenol ≥97%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

L12031.03 L12031.06
L12031.03EA 23 EUR
L12031.03 L12031.06
3,4-Dichlorothiophenol ≥97%
3,4-Dichlorothiophenol
Formule: C₆H₄Cl₂S
Kookpunt: 119…120 °C (10 mmHg)
Smeltpunt: 87…89 °C
Dichtheid: 1,407
Vlampunt: >110 °C (230 °F)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00004840
CAS nummer: 5858-17-3
EINECS: 227-485-3

Order Now

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR