1,2-Ethanedithiol ≥98%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

L12865.14 L12865.22 L12865.36
L12865.14EA 26.5 EUR
L12865.14 L12865.22 L12865.36
1,2-Ethanedithiol ≥98%
1,2-Ethanedithiol

Purity: 98+% CAS No.: 540-63-6 Molecular Formula: C2H6S2 Formula Weight: 94.20 Flash Point: 45°(113°F) Refractive Index: 1.5580 Odour: Stench Melting Point: -41° Boiling Point: 144-146° Beilstein Reference: 505946 Merck Reference: 14,3725 UN No.: 3071 MDL No.: MFCD00004892

Formule: C₂H₆S₂
Kookpunt: 144…146 °C
Smeltpunt: –41 °C
Dichtheid: 1,123
Vlampunt: 45 °C (113 °F)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00004892
CAS nummer: 540-63-6
EINECS: 208-752-3
UN: 3071
ADR: 6.1,II
Merck Index: 14,3725

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR