(4-Iodophenyl)acetic acid ≥97%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

L13345.03 L13345.06
L13345.03EA 37.8 EUR
L13345.03 L13345.06
(4-Iodophenyl)acetic acid ≥97%
(4-Iodophenyl)acetic acid

Purity: 97% CAS No.: 1798-06-7 Molecular Formula: C8H7IO2 Formula Weight: 262.05 Melting Point: 136-140° Beilstein Reference: 1940558

Formule: (C₆H₄I)CH₂COOH
Smeltpunt: 136…140 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00082985
CAS nummer: 1798-06-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR