Magnesium sulphate hydrate, (dried basis) contains ca 1-2 mol water of hydration

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

L13739.22 L13739.36 L13739.0E
L13739.22EA 21.8 EUR
L13739.22 L13739.36 L13739.0E
Magnesium sulphate hydrate, (dried basis) contains ca 1-2 mol water of hydration
Magnesium sulphate hydrate

CAS No.: 22189-08-8 Molecular Formula: MgSO4.xH2O Formula Weight: 120.37(anhy) Merck Reference: 14,5691 MDL No.: MFCD00149787

Formule: MgSO₄·nH₂O
MW: 120.37 g/mol (anhy)
Dichtheid: 2.45 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00011110
CAS nummer: 22189-08-8
EINECS: 231-298-2
Merck Index: 14,5691

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR