Pyrrole-2-carboxylic acid ≥99%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

L13756.03 L13756.06 L13756.14
L13756.03EA 42.6 EUR
L13756.03 L13756.06 L13756.14
Pyrrole-2-carboxylic acid ≥99%
Pyrrole-2-carboxylic acid

Purity: 99% CAS No.: 634-97-9 Molecular Formula: C5H5NO2 Formula Weight: 111.10 Melting Point: ca 205° subl. Beilstein Reference: 80825

Formule: C₅H₅NO₂
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00005219
CAS nummer: 634-97-9
EINECS: 211-221-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR