Tetrapentylammonium iodide ≥98%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

L14088.22 L14088.06 L14088.14
L14088.22EA 234 EUR
L14088.22 L14088.06 L14088.14
Tetrapentylammonium iodide ≥98%
Tetrapentylammonium iodide

Purity: 98% CAS No.: 2498-20-6 Molecular Formula: C20H44IN Formula Weight: 425.49 Melting Point: 135-139° Beilstein Reference: 3917412

Formule: C₂₀H₄₄IN
Smeltpunt: 135…139 °C
Dichtheid: 1,18 g/cm³ (25 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00041980
CAS nummer: 2498-20-6
EINECS: 219-688-0

Order Now

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR