4-Aminobenzamidine dihydrochloride 97%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

L14089.03 L14089.06
L14089.03EA 32.7 EUR
L14089.03 L14089.06
4-Aminobenzamidine dihydrochloride 97%
4-Aminobenzamidine dihydrochloride

Purity: 97% CAS No.: 2498-50-2 Molecular Formula: C7H9N3.2HCl Formula Weight: 208.09 Storage Temperature: 2°C to 8°C Melting Point: 300° Beilstein Reference: 3692927 MDL No.: MFCD00013001

Formule: C₇H₁₁Cl₂N₃
Smeltpunt: >300 °C
MDL Nummer: MFCD00013001
CAS nummer: 2498-50-2
EINECS: 219-692-2

Order Now

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR