1,2-Diethoxyethaan ≥98%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

Synoniemen: Diethyl glycol, Ethylene glycol diethyl ether

L14283.AE L14283.AP
L14283.AEEA 39.3 EUR
L14283.AE L14283.AP
1,2-Diethoxyethaan ≥98%
1,2-Diethoxyethaan

Purity: 98% CAS No.: 629-14-1 Molecular Formula: C6H14O2 Formula Weight: 118.18 Flash Point: 19°(66°F) Refractive Index: 1.3920 Melting Point: -74° Boiling Point: 120-121° Beilstein Reference: 1732917 UN No.: 1153 MDL No.: MFCD00009253

Formule: C₆H₁₄O₂
MW: 118.18 g/mol
Kookpunt: 120…121 °C
Smeltpunt: –74 °C
Dichtheid: 0.842
Vlampunt: 19 °C (66 °F)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00009253
CAS nummer: 629-14-1
EINECS: 211-076-1
UN: 1153
ADR: 3 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR