2-Chloro-6-fluorobenzaldehyde oxime ≥97%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

L14719.03 L14719.06
L14719.03EA 16.6 EUR
L14719.03 L14719.06
2-Chloro-6-fluorobenzaldehyde oxime ≥97%
2-Chloro-6-fluorobenzaldehyde oxime

Purity: 97% CAS No.: 443-33-4 Molecular Formula: C7H5ClFNO Formula Weight: 173.58 Melting Point: 125-127° Beilstein Reference: 3247886

Formule: C₇H₅ClFNO
Smeltpunt: 125…127 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00837704
CAS nummer: 443-33-4
EINECS: 207-135-6

Order Now

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR