Tripotassium orthophosphate ≥97%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

Synoniemen: Potassium phosphate tribasic, Potassium phosphate

L15168.0P L15168.22 L15168.36
L15168.0PEA 134 EUR
L15168.0P L15168.22 L15168.36
Tripotassium orthophosphate ≥97%
Tripotassium orthophosphate
Formule: K₃O₄P
MW: 212.27 g/mol
Smeltpunt: 1340
Dichtheid: 2.564 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00036295
CAS nummer: 7778-53-2
EINECS: 231-907-1
Merck Index: 12,07830

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR