Methylenediphenyl-4,4'-diisocyanate 98%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

L15740.14 L15740.22 L15740.36
L15740.14EA 24.2 EUR
L15740.14 L15740.22 L15740.36
Methylenediphenyl-4,4'-diisocyanate 98%
Methylenediphenyl-4,4'-diisocyanate

Purity: 98% CAS No.: 101-68-8 Molecular Formula: C15H10N2O2 Formula Weight: 250.26 Storage Temperature: -30°C to -10°C Flash Point: 196°(385°F) Odour: Weak, characteristic Melting Point: 41-44° Boiling Point: 194°/5mm Beilstein Reference: 797662 UN No.: UN2811 MDL No.: MFCD00036131

Formule: C₁₅H₁₀N₂O₂
Kookpunt: 194 °C (5 mmHg)
Smeltpunt: 41…44 °C
Dichtheid: 1,239
Vlampunt: 196 °C (385 °F)
Bewaar temperatuur: Vriezer
MDL Nummer: MFCD00036131
CAS nummer: 101-68-8
EINECS: 202-966-0
UN: 2811
ADR: 6.1,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR