1,6-Dihydroxynaphthalene ≥97%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

L15801.14 L15801.22 L15801.36
L15801.14EA 46.5 EUR
L15801.14 L15801.22 L15801.36
1,6-Dihydroxynaphthalene ≥97%
1,6-Dihydroxynaphthalene

Purity: 97+% CAS No.: 575-44-0 Molecular Formula: C10H8O2 Formula Weight: 160.17 Melting Point: 136-140° Beilstein Reference: 1939032

Formule: C₁₀H₈O₂
Kookpunt: 375 °C (1013 hPa)
Smeltpunt: 136…140 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00003981
CAS nummer: 575-44-0
EINECS: 209-386-7

Order Now

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR