1,4-Dibutylbenzene ≥97%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

L16396.03 L16396.09
L16396.03EA 39 EUR
L16396.03 L16396.09
1,4-Dibutylbenzene ≥97%
1,4-Dibutylbenzene

Purity: 97+% CAS No.: 1571-86-4 Molecular Formula: C14H22 Formula Weight: 190.33 Flash Point: 110°(230°F) Refractive Index: 1.4890 Melting Point: -31° Boiling Point: 227° Beilstein Reference: 1858055 MDL No.: MFCD01076591

Formule: C₁₄H₂₂
MW: 190.33
Kookpunt: 227 °C
Smeltpunt: –31 °C
Dichtheid: 0.856
Vlampunt: 110 °C (230 °F)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD01076591
CAS nummer: 1571-86-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR