3,3,3-Trifluoro-1-propanol ≥97%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

L16879.03 L16879.06
L16879.03EA 87 EUR
L16879.03 L16879.06
3,3,3-Trifluoro-1-propanol ≥97%
3,3,3-Trifluoro-1-propanol

Purity: 97% CAS No.: 2240-88-2 Molecular Formula: CF3CH2CH2OH Formula Weight: 114.07 Flash Point: 37°(98°F) Refractive Index: 1.3200 Boiling Point: 100-101° Beilstein Reference: 1734777 UN No.: 1987 MDL No.: MFCD00190649

Formule: C₃H₅F₃O
Kookpunt: 100…101 °C
Dichtheid: 1,294
Vlampunt: 37 °C (98 °F)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00190649
CAS nummer: 2240-88-2
UN: 1987
ADR: 3,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR