Ammoniumwaterstofdifluoride 95% remainder mainly ammonium fluoride

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

L17237.22 L17237.36 L17237.0E L17237.C4
L17237.22EA 29.7 EUR
L17237.22 L17237.36 L17237.0E L17237.C4
Ammoniumwaterstofdifluoride 95% remainder mainly ammonium fluoride
Ammoniumwaterstofdifluoride

Purity: 95% CAS No.: 1341-49-7 Molecular Formula: H5F2N Formula Weight: 57.04 Odour: Ammonia-like pH: 2-3 Melting Point: 120-122° Boiling Point: 238° dec. Merck Reference: 14,498 UN No.: 1727 MDL No.: MFCD00012018

Formule: NH₄HF₂
Kookpunt: 238 °C (ontbinding)
Smeltpunt: 120…122 °C
Dichtheid: 1,500
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00012018
CAS nummer: 1341-49-7
EINECS: 215-676-4
UN: 1727
ADR: 8,II
Merck Index: 14,498

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR