N-(2-Pyridyl)bis(trifluoromethanesulphonimide) 98%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

L17433.06
L17433.06EA 164 EUR
L17433.06
N-(2-Pyridyl)bis(trifluoromethanesulphonimide) 98%
N-(2-Pyridyl)bis(trifluoromethanesulphonimide)

Purity: 98% CAS No.: 145100-50-1 Molecular Formula: C7H4F6N2O4S2 Formula Weight: 358.24 Storage Temperature: 2°C to 8°C Flash Point: 110°(230°F) Melting Point: 37-41° Boiling Point: 97°/2mm Beilstein Reference: 5832565 MDL No.: MFCD00191834

Formule: C₇H₄F₆N₂O₄S₂
Kookpunt: 97 °C (2 mmHg)
Smeltpunt: 37…41 °C
Vlampunt: >110 °C (230 °F)
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00191834
CAS nummer: 145100-50-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR