6-Bromoindole ≥98%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

L17703.03 L17703.MD
L17703.03EA 148 EUR
L17703.03 L17703.MD
6-Bromoindole ≥98%
6-Bromoindole

Purity: 98% CAS No.: 52415-29-9 Molecular Formula: C8H6BrN Formula Weight: 196.05 Melting Point: 91-93° Boiling Point: 70-75°/0.01mm MDL No.: MFCD00238550

Formule: C₈H₆BrN
Kookpunt: 70…75 °C (0,01 mmHg)
Smeltpunt: 91…93 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00238550
CAS nummer: 52415-29-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR