5-(4-Fluorophenyl)valeric acid 97%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

L17935.03 L17935.06 L17935.MD
L17935.03EA 86.3 EUR
L17935.03 L17935.06 L17935.MD
5-(4-Fluorophenyl)valeric acid 97%
5-(4-Fluorophenyl)valeric acid

Purity: 97% CAS No.: 24484-22-8 Molecular Formula: C11H13FO2 Formula Weight: 196.22 Melting Point: 73-75° Beilstein Reference: 7918899 MDL No.: MFCD00800242

Formule: C₁₁H₁₃FO₂
Smeltpunt: 73…75 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00800242
CAS nummer: 24484-22-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR